Pyridazine 98+%
Supplier: Thermo Scientific Chemicals
Warning
|
A13266-06
A13266-14
CAAAA13266-06EA
91.8
CAD
CAAAA13266-06
CAAAA13266-14
Pyridazine 98+%
Pyridazine
MDL: MFCD00006463
Beilstein: 103906
Miscible with water, benzene, DMF. Soluble in alcohol, ether
Purity: 98+%
CAS No.: 289-80-5
Molecular Formula: C4H4N2
Formula Weight: 80.09
Flash Point: 85°(185°F)
Refractive Index: 1.5240
Odor: Unpleasant
Melting Point: -8°
Boiling Point: 87°/14mm
Solubility: Miscible with water, benzene, DMF. Soluble in alcohol, ether
Beilstein Reference: 103906
Merck Reference: 14,7969
MDL No.: MFCD00006463
Formula:
C₄H₄N₂ Boiling Pt: 87 °C/14 mm Melting Pt: –8 °C Density: 1.105 Flash Pt: 85 °C(185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006463 CAS Number: 289-80-5 EINECS: 206-025-5 Merck Index: 14,7969 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...