Methyl stearate 99%

Supplier: Thermo Scientific Chemicals

A13265-06 A13265-14
CAAAA13265-06EA 124.19 CAD
CAAAA13265-06 CAAAA13265-14
Methyl stearate 99%
Methyl stearate

Purity: 99% CAS No.: 112-61-8 Molecular Formula: CH3(CH2)16CO2CH3 Formula Weight: 298.51 Flash Point: 110°(230°F) Physical Form: Cryst powder Melting Point: 38-42° Boiling Point: 180-182/4mm Beilstein Reference: 1786213 Merck Reference: 14,8804 MDL No.: MFCD00009005

Formula: CH₃(CH₂)₁₆CO₂CH₃
Boiling Pt: 180…182/4 mm
Melting Pt: 38…42 °C
Density: 1.007 g/cm³ (20 °C)
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009005
CAS Number: 112-61-8
EINECS: 203-990-4
Merck Index: 14,8804

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR