3-Mercaptopropionic acid 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13261-22
A13261-36
CAAAA13261-22EA
45.35
CAD
CAAAA13261-22
CAAAA13261-36
3-Mercaptopropionic acid 99%
3-Mercaptopropionic acid
Purity: 99%
CAS No.: 107-96-0
Molecular Formula: C3H6O2S
Formula Weight: 106.14
Flash Point: 93°(199°F)
Refractive Index: 1.4920
Melting Point: 17-19°
Boiling Point: 110-111°/15mm
Beilstein Reference: 773807
UN No.: 2922
MDL No.: MFCD00004897
Formula:
C₃H₆O₂S Boiling Pt: 110…111 °C/15 mm Melting Pt: 17…19 °C Density: 1.220 Flash Pt: 93 °C (199 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004897 CAS Number: 107-96-0 EINECS: 203-537-0 UN: 2922 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...