3-Chlorobenzaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13254-14
A13254-22
A13254-36
CAAAA13254-14EA
89.74
CAD
CAAAA13254-14
CAAAA13254-22
CAAAA13254-36
3-Chlorobenzaldehyde 97%
3-Chlorobenzaldehyde
Purity: 97%
CAS No.: 587-04-2
Molecular Formula: C7H5ClO
Formula Weight: 140.57
Flash Point: 88°(190°F)
Refractive Index: 1.5640
Odor: Characteristic
Melting Point: 16-18°
Boiling Point: 213-214°
Beilstein Reference: 507098
MDL No.: MFCD00003350
Formula:
C₇H₅ClO Boiling Pt: 213…214 °C Melting Pt: 16…18 °C Density: 1,239 Flash Pt: 88 °C(190 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003350 CAS Number: 587-04-2 EINECS: 209-596-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...