1,3-Difluoro-4-nitrobenzene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13249-14
A13249-22
A13249-36
CAAAA13249-14EA
48.66
CAD
CAAAA13249-14
CAAAA13249-22
CAAAA13249-36
1,3-Difluoro-4-nitrobenzene 98%
1,3-Difluoro-4-nitrobenzene
Purity: 98%
CAS No.: 446-35-5
Molecular Formula: C6H3F2NO2
Formula Weight: 159.09
Flash Point: 90°(194°F)
Refractive Index: 1.5130
Melting Point: 9-10°
Boiling Point: 203-204°
Beilstein Reference: 2048034
UN No.: 2810
MDL No.: MFCD00007050
Formula:
C₆H₃F₂NO₂ Boiling Pt: 203…204 °C Melting Pt: 9…10 °C Density: 1.450 Flash Pt: 90 °C (194 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007050 CAS Number: 446-35-5 EINECS: 207-167-0 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...