Propiolic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13245-06
A13245-14
A13245-22
CAAAA13245-06EA
71.57
CAD
CAAAA13245-06
CAAAA13245-14
CAAAA13245-22
Propiolic acid 98+%
Propiolic acid
Purity: 98+%
CAS No.: 471-25-0
Molecular Formula: HC≡CCO2H
Formula Weight: 70.05
Storage Temperature: 2°C to 8°C
Flash Point: 58°(136°F)
Refractive Index: 1.4330
Melting Point: 18°
Boiling Point: 102°/200mm
Beilstein Reference: 878176
Merck Reference: 14,7824
UN No.: 2920
MDL No.: MFCD00004360
Formula:
C₃H₂O₂ Boiling Pt: 102 °C/200 mm Melting Pt: 18 °C Density: 1.136 Flash Pt: 58 °C (136 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00004360 CAS Number: 471-25-0 EINECS: 207-437-8 UN: 2920 ADR: 8,II Merck Index: 14,7824 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...