L-Phenylalanine 99%

Supplier: Thermo Scientific Chemicals

Synonyms: H-L-Phe-OH, Phenylalanine, H-Phe-OH, L-α-Phenylalanine, (S)-(-)-Phenylalanine, 3-Phenyl-L-alanine, 3-Phenyl-L(-)-alanine

A13238-14 A13238-22 A13238-36 A13238-0B
CAAAAA13238-14EA 58.62 CAD
CAAAAA13238-14 CAAAAA13238-22 CAAAAA13238-36 CAAAA13238-0B CAAAA13238-18
L-Phenylalanine 99%
L-α-Phenylalanine

Purity: 99% CAS No.: 63-91-2 Molecular Formula: C9H11NO2 Formula Weight: 165.19 Physical Form: Cryst powder Appearance: White Melting Point: ca 275° dec. Beilstein Reference: 1910408 Merck Reference: 14,7271 Optical Rotation: -34° (c=2 in water) MDL No.: MFCD00064227

Formula: C₉H₁₁NO₂
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: ∼ 275 °C (dec.)
Density: 0.754 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064227
CAS Number: 63-91-2
EINECS: 200-568-1
Merck Index: 14,7271

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR