3-Chlorobenzonitrile 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: m-Chlorobenzonitrile

A13237-30 A13237-09 A13237-18
CAAAAA13237-30EA 910.08 CAD
CAAAAA13237-30 CAAAA13237-09 CAAAA13237-18
3-Chlorobenzonitrile 99%
3-Chlorobenzonitrile

Purity: 99% CAS No.: 766-84-7 Molecular Formula: C7H4ClN Formula Weight: 137.57 Flash Point: 97°(206°F) Odor: Almond-like Boiling Point: 94°/10mm Beilstein Reference: 1906694 UN No.: 3439 MDL No.: MFCD00001798

Formula: C₇H₄ClN
MW: 137.57 g/mol
Boiling Pt: 203
Melting Pt: 39
Density: 1.14
Flash Pt: 97 °C (206 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001798
CAS Number: 766-84-7
EINECS: 212-172-6
UN: 3439

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR