4-Iodo-o-xylene 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,4-Dimethyliodobenzene, 1,2-Dimethyl-4-iodobenzene, 3,4-Dimethyl-1-iodobenzene, 4-Iodo-1,2-dimethylbenzene, 1-Iodo-3,4-dimethylbenzene

A13232-30 A13232-09 A13232-18
CAAAAA13232-30EA 913.24 CAD
CAAAAA13232-30 CAAAA13232-09 CAAAA13232-18
4-Iodo-o-xylene 98+%
4-Iodo-o-xylene

Purity: 98+% CAS No.: 31599-61-8 Molecular Formula: C8H9I Formula Weight: 232.07 Flash Point: 110°(230°F) Refractive Index: 1.6020 Boiling Point: 231-233° Beilstein Reference: 1905729 MDL No.: MFCD00040989

Formula: C₈H₉I
Boiling Pt: 231…233 °C
Density: 1.633
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040989
CAS Number: 31599-61-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR