(4-Tolyloxy)acetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

A13230-09 A13230-18 A13230-30
CAAAA13230-09EA 49.77 CAD
CAAAA13230-09 CAAAA13230-18 CAAAA13230-30
(4-Tolyloxy)acetic acid 98%
(4-Tolyloxy)acetic acid

Purity: 98% CAS No.: 940-64-7 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 137-142° Beilstein Reference: 2047591

Formula: (C₆H₄(CH₃))OCH₂COOH
Melting Pt: 137…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00014365
CAS Number: 940-64-7
EINECS: 213-374-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR