4-tert-Butylbenzhydrazide 98+%
Supplier: Thermo Scientific Chemicals
Synonyms:
4-tert-Butylbenzoic hydrazide, 4-(tert-Butyl)benzohydrazide, 4-tert-Butylbenzene-1-carbohydrazide, 4-tert-Butylbenzoic acid hydrazide, 4-tert-Butylbenzohydrazide
A13224-14
A13224-22
CAAAA13224-14EA
142.52
CAD
CAAAA13224-14
CAAAA13224-22
CAAAA13224-36
4-tert-Butylbenzhydrazide 98+%
4-tert-Butylbenzhydrazide
Purity: 98+%
CAS No.: 43100-38-5
Molecular Formula: C11H16N2O
Formula Weight: 192.26
Melting Point: 127-131°
Beilstein Reference: 1104663
Formula:
C₁₁H₁₆N₂O Melting Pt: 127…131 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014763 CAS Number: 43100-38-5 EINECS: 256-090-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...