3-Aminopyridine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13216-0B
A13216-18
A13216-30
CAAAA13216-0BEA
628.84
CAD
CAAAA13216-0B
CAAAA13216-18
CAAAA13216-30
3-Aminopyridine 99%
3-Aminopyridine
Purity: 99%
CAS No.: 462-08-8
Molecular Formula: C5H6N2
Formula Weight: 94.12
Flash Point: 88°(190°F)
Odor: Unpleasant
pH: 10
Melting Point: 58-63°
Boiling Point: 250-252°
Beilstein Reference: 105692
Merck Reference: 14,473
UN No.: 2671
MDL No.: MFCD00006400
Formula:
C₅H₆N₂ Boiling Pt: 250…252 °C Melting Pt: 58…63 °C Flash Pt: 88 °C (190 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006400 CAS Number: 462-08-8 EINECS: 207-322-2 UN: 2671 ADR: 6.1,II Merck Index: 14,473 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...