4'-Aminoacetophenone 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(4-Aminophenyl)ethanone, p-Aminoacetophenone

A13213-14 A13213-0B A13213-18 A13213-30
CAAAAA13213-14EA 52.93 CAD
CAAAAA13213-14 CAAAA13213-0B CAAAA13213-18 CAAAA13213-30
4'-Aminoacetophenone 99%
4'-Aminoacetophenone

Purity: 99% CAS No.: 99-92-3 Molecular Formula: C8H9NO Formula Weight: 135.17 Melting Point: 103-108° Boiling Point: 292-294° Merck Reference: 14,413 MDL No.: MFCD00007896

Formula: C₈H₉NO
Boiling Pt: 292…294 °C
Melting Pt: 103…108 °C
Density: 1.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007896
CAS Number: 99-92-3
EINECS: 202-801-2
Merck Index: 14,413

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR