2'-Nitroacetophenone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: o-Nitroacetophenone, 1-(2-Nitrophenyl)ethanone

A13208-09 A13208-18
CAAAA13208-09EA 145.83 CAD
CAAAA13208-09 CAAAA13208-18
2'-Nitroacetophenone 97%
2'-Nitroacetophenone

Purity: 97% CAS No.: 577-59-3 Molecular Formula: C8H7NO3 Formula Weight: 165.15 Flash Point: 110°(230°F) Refractive Index: 1.5515 Odor: Characteristic Melting Point: 25-28° Boiling Point: 158-160°/16mm Beilstein Reference: 1102322 MDL No.: MFCD00007144

Formula: C₈H₇NO₃
MW: 165.15 g/mol
Boiling Pt: 158…160 °C (16 mmHg)
Melting Pt: 25…28 °C
Density: 1.24
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007144
CAS Number: 577-59-3
EINECS: 209-414-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR