2,4-Dichlorobenzylamine 98%

Supplier: Thermo Scientific Chemicals
Danger

A13207-14 A13207-06
CAAAAA13207-14EA 195.92 CAD
CAAAAA13207-14 CAAAA13207-03 CAAAA13207-06
2,4-Dichlorobenzylamine 98%
2,4-Dichlorobenzylamine

Purity: 98% CAS No.: 95-00-1 Molecular Formula: C7H7Cl2N Formula Weight: 176.05 Flash Point: 110°(230°F) Refractive Index: 1.5780 Odor: Amine-like Boiling Point: 258-260° Beilstein Reference: 775667 UN No.: 2735 MDL No.: MFCD00008109

Formula: C₇H₇Cl₂N
Boiling Pt: 258…260 °C
Density: 1.320
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008109
CAS Number: 95-00-1
EINECS: 202-382-6
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR