Propiononitrile 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Propionitrile

A13203-AK A13203-AU
CAAAA13203-AKEA 91.01 CAD
CAAAA13203-AK CAAAA13203-AU
Propiononitrile 99%
Propiononitrile

Purity: 99% CAS No.: 107-12-0 Molecular Formula: CH3CH2CN Formula Weight: 55.08 Flash Point: 6°(42°F) Refractive Index: 1.3660 Odor: Pleasant Melting Point: -92° Boiling Point: 97-98° Beilstein Reference: 773680 Merck Reference: 14,7827 UN No.: 2404 MDL No.: MFCD00001948

Formula: C₃H₅N
MW: 55.08 g/mol
Boiling Pt: 97…98 °C
Melting Pt: –92 °C
Density: 0.781
Flash Pt: 6 °C (42 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001948
CAS Number: 107-12-0
EINECS: 203-464-4
UN: 2404
ADR: 3,II
Merck Index: 14,7827

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR