2,3-Dimethyl-2-butene 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Tetramethylethylene
A13201-06
A13201-14
CAAAA13201-06EA
64.15
CAD
CAAAA13201-06
CAAAA13201-14
CAAAA13201-22
2,3-Dimethyl-2-butene 97%
2,3-Dimethyl-2-butene
Purity: 97%
CAS No.: 563-79-1
Molecular Formula: C6H12
Formula Weight: 84.16
Flash Point: -17°(1°F)
Refractive Index: 1.4120
Odor: Mild
Melting Point: -75°
Boiling Point: 72-73°
Beilstein Reference: 1361357
UN No.: 3295
MDL No.: MFCD00008897
Formula:
C₆H₁₂ MW: 84.16 g/mol Boiling Pt: 72…73 °C Melting Pt: –75 °C Density: 0.705 Flash Pt: –17 °C (1 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008897 CAS Number: 563-79-1 EINECS: 209-263-8 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...