2-Methylbenzoxazole 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Methyl-1,3-benzoxazole, 2-Methylbenzo[d]oxazole
A13198-14
A13198-22
CAAAA13198-14EA
89.9
CAD
CAAAA13198-14
CAAAA13198-22
2-Methylbenzoxazole 98%
2-Methylbenzoxazole
Purity: 98%
CAS No.: 95-21-6
Molecular Formula: C8H7NO
Formula Weight: 133.15
Flash Point: 85°(185°F)
Refractive Index: 1.5500
Melting Point: 2-4°
Boiling Point: 200-201°
Beilstein Reference: 112297
MDL No.: MFCD00005771
Formula:
C₈H₇NO MW: 133.15 g/mol Boiling Pt: 200 Melting Pt: 2…4 °C Density: 1.120 Flash Pt: 85 °C (185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005771 CAS Number: 95-21-6 EINECS: 202-399-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...