4-Bromobenzyl alcohol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(4-Bromophenyl)methanol
A13197-09
A13197-18
CAAAA13197-09EA
89.59
CAD
CAAAA13197-09
CAAAA13197-18
4-Bromobenzyl alcohol 99%
4-Bromobenzyl alcohol
Purity: 99%
CAS No.: 873-75-6
Molecular Formula: C7H7BrO
Formula Weight: 187.04
Melting Point: 76-79°
Beilstein Reference: 1931620
Formula:
C₇H₇BrO Boiling Pt: 268 °C (1013 hPa) Melting Pt: 76…79 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004650 CAS Number: 873-75-6 EINECS: 212-851-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...