2-Methoxynicotinic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Methoxy-3-pyridinecarboxylic acid, 2-Methoxypyridine-3-carboxylic acid

A13193-03 A13193-06
CAAAA13193-03EA 29.07 CAD
CAAAA13193-03 CAAAA13193-06
2-Methoxynicotinic acid 98%
2-Methoxynicotinic acid

Purity: 98% CAS No.: 16498-81-0 Molecular Formula: C7H7NO3 Formula Weight: 153.14 Melting Point: 147-151° MDL No.: MFCD00085110

Formula: C₇H₇NO₃
MW: 153.14 g/mol
Melting Pt: 147…151 °C
Storage Temperature: Ambient
MDL Number: MFCD00085110
CAS Number: 16498-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR