1,3-Dichloro-5-iodobenzene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13192-06
A13192-14
A13192-22
CAAAA13192-06EA
66.68
CAD
CAAAA13192-06
CAAAA13192-14
CAAAA13192-22
1,3-Dichloro-5-iodobenzene 99%
1,3-Dichloro-5-iodobenzene
Purity: 99%
CAS No.: 3032-81-3
Molecular Formula: C6H3Cl2I
Formula Weight: 272.90
Melting Point: 55-59°
Boiling Point: 127°/20mm
MDL No.: MFCD00001047
Formula:
C₆H₃Cl₂I Boiling Pt: 127 °C/20 mm Melting Pt: 55…59 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001047 CAS Number: 3032-81-3 EINECS: 221-216-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...