1,2-Epoxycyclohexane 98+%

Supplier: Thermo Scientific Chemicals
Danger

A13185-0E A13185-22 A13185-36
CAAAA13185-0EEA 970.12 CAD
CAAAA13185-0E CAAAA13185-22 CAAAA13185-36
1,2-Epoxycyclohexane 98+%
1,2-Epoxycyclohexane

MDL: MFCD00005162 Beilstein: 383568 Insoluble in water


Purity: 98+% CAS No.: 286-20-4 Molecular Formula: C6H10O Formula Weight: 98.15 Flash Point: 24°(75°F) Refractive Index: 1.4520 Melting Point: -40° Boiling Point: 129-130° Solubility: Insoluble in water Beilstein Reference: 383568 UN No.: 2920 MDL No.: MFCD00005162

Formula: C₆H₁₀O
Boiling Pt: 129…130 °C
Melting Pt: –40 °C
Density: 0.972
Flash Pt: 24 °C (75 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005162
CAS Number: 286-20-4
EINECS: 206-007-7
UN: 2920
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR