2-Aminobenzyl alcohol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13180-09
A13180-18
A13180-30
CAAAA13180-09EA
52.77
CAD
CAAAA13180-09
CAAAA13180-18
CAAAA13180-30
2-Aminobenzyl alcohol 98%
2-Aminobenzyl alcohol
Purity: 98%
CAS No.: 5344-90-1
Molecular Formula: C7H9NO
Formula Weight: 123.16
Storage Temperature: 2°C to 8°C
Melting Point: 81-84°
Boiling Point: 160°/10mm
Beilstein Reference: 1072211
MDL No.: MFCD00007749
Formula:
C₇H₉NO Boiling Pt: 160 °C/10 mm Melting Pt: 81…84 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00007749 CAS Number: 5344-90-1 EINECS: 226-293-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...