(2-Chlorophenyl)acetonitrile 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Chlorobenzyl cyanide, 2-(2-Chlorophenyl)acetonitrile

A13176-09 A13176-18
CAAAA13176-09EA 44.71 CAD
CAAAA13176-09 CAAAA13176-18
(2-Chlorophenyl)acetonitrile 98+%
(2-Chlorophenyl)acetonitrile

Purity: 98+% CAS No.: 2856-63-5 Molecular Formula: C8H6ClN Formula Weight: 151.60 Flash Point: 110°(230°F) Refractive Index: 1.5440 Melting Point: 21-24° Boiling Point: 240-242° Beilstein Reference: 2043683 UN No.: 3439 MDL No.: MFCD00001898

Formula: C₈H₆ClN
Boiling Pt: 240…242 °C
Melting Pt: 21…24 °C
Density: 1 g/cm³ (20 °C)
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001898
CAS Number: 2856-63-5
EINECS: 220-669-4
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR