4-Methylpyrimidine 98%

Supplier: Thermo Scientific Chemicals
Danger

A13170-06 A13170-14
CAAAA13170-06EA 129.4 CAD
CAAAA13170-06 CAAAA13170-14 CAAAA13170-22
4-Methylpyrimidine 98%
4-Methylpyrimidine

Purity: 98% CAS No.: 3438-46-8 Molecular Formula: C5H6N2 Formula Weight: 94.12 Flash Point: 40°(104°F) Refractive Index: 1.4960 Boiling Point: 141-142° Beilstein Reference: 105781 UN No.: 1993 MDL No.: MFCD00006115

Formula: C₅H₆N₂
MW: 94.12
Boiling Pt: 141…142 °C
Melting Pt: 32
Density: 1.02
Flash Pt: 40 °C (104 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006115
CAS Number: 3438-46-8
EINECS: 222-344-2
UN: 1993
ADR: 3 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR