6-Chloro-2-hydroxypyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Chloro-6-pyridinol, 6-Chloro-2-pyridinol, 2-Chloro-6-hydroxypyridine, 6-Chloropyridin-2(1H)-one
A13164-14
A13164-22
CAAAA13164-14EA
94.17
CAD
CAAAA13164-14
CAAAA13164-22
6-Chloro-2-hydroxypyridine 98%
6-Chloro-2-hydroxypyridine
Purity: 98%
CAS No.: 16879-02-0
Molecular Formula: C5H4ClNO
Formula Weight: 129.55
Flash Point: 163°(325°F)
Melting Point: 129-131°
Beilstein Reference: 108665
MDL No.: MFCD00006278
Formula:
C₅H₄ClNO MW: 129.55 g/mol Melting Pt: 129…131 °C Flash Pt: 163 °C (325 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006278 CAS Number: 16879-02-0 EINECS: 240-909-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...