p-Benzoquinone 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
p-Quinone, Quinone, 1,4-Cyclohexadiene-3,6-dione, 1,4-Benzoquinone
A13162-22
A13162-0B
A13162-0I
A13162-30
CAAAAA13162-22EA
56.56
CAD
CAAAAA13162-22
CAAAA13162-0B
CAAAA13162-0I
CAAAA13162-30
p-Benzoquinone 98+%
p-Benzoquinone
Purity: 98+%
CAS No.: 106-51-4
Molecular Formula: C6H4O2
Formula Weight: 108.10
Flash Point: 77°(171°F)
Physical Form: Powder
Density: 1.318
Odor: Pungent
Melting Point: 112-115°
Boiling Point: ca 180° subl.
Beilstein Reference: 773967
Merck Reference: 14,8074
UN No.: 2587
MDL No.: MFCD00001591
Formula:
C₆H₄O₂ Boiling Pt: ∼ 180 °C (subl.) Melting Pt: 112…115 °C Density: 1.318 Flash Pt: 77 °C (171 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001591 CAS Number: 106-51-4 EINECS: 203-405-2 UN: 2587 ADR: 6.1,II Merck Index: 14,8074 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...