4-Bromoisophthalic acid 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13152-06
A13152-14
A13152-22
CAAAA13152-06EA
111.86
CAD
CAAAA13152-06
CAAAA13152-14
CAAAA13152-22
4-Bromoisophthalic acid 96%
4-Bromoisophthalic acid
Purity: 96%
CAS No.: 6939-93-1
Molecular Formula: C8H5BrO4
Formula Weight: 245.03
Melting Point: 296-302°
Beilstein Reference: 1959392
Formula:
C₈H₅BrO₄ MW: 245.03 g/mol Melting Pt: 296…302 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002404 CAS Number: 6939-93-1 EINECS: 230-078-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...