2-Hydroxyisobutyric acid, (max. 2% H₂O) 99% (dry weight)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Hydroxy-2-methylpropionic acid, α-Hydroxyisobutyric acid, 2-Hydroxy-2-methylpropanoic acid

A13146-14 A13146-22 A13146-36
CAAAA13146-14EA 56.56 CAD
CAAAA13146-14 CAAAA13146-22 CAAAA13146-36
2-Hydroxyisobutyric acid, (max. 2% H₂O) 99% (dry weight)
2-Hydroxyisobutyric acid

Purity: 99% CAS No.: 594-61-6 Molecular Formula: C4H8O3 Formula Weight: 104.11 Melting Point: 77-80° Boiling Point: 114°/12mm MDL No.: MFCD00004459

Formula: C₄H₈O₃
MW: 104.11 g/mol
Boiling Pt: 212
Melting Pt: 77
Density: 1.24
Storage Temperature: Ambient
MDL Number: MFCD00004459
CAS Number: 594-61-6
EINECS: 209-848-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR