DL-Proline 99%

Supplier: Thermo Scientific Chemicals

Synonyms: (±)-Proline, H-DL-Pro-OH

A13137-06 A13137-14 A13137-22
CAAAA13137-06EA 67.31 CAD
CAAAA13137-06 CAAAA13137-14 CAAAA13137-22
DL-Proline 99%
DL-Proline

Purity: 99% CAS No.: 609-36-9 Molecular Formula: C5H9NO2 Formula Weight: 115.13 Melting Point: ca 209° dec. Beilstein Reference: 80809 Merck Reference: 14,7780 MDL No.: MFCD00005250

Formula: C₅H₉NO₂
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: ∼ 209 °C (dec.)
Density: 1.38 g/cm³ (22.9 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005250
CAS Number: 609-36-9
EINECS: 210-189-3
Merck Index: 14,7780

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR