2'-Hydroxy-5'-methylacetophenone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-(2-Hydroxy-5-methylphenyl)ethanone
A13136-22
A13136-06
A13136-14
CAAAAA13136-22EA
808.96
CAD
CAAAAA13136-22
CAAAA13136-06
CAAAA13136-14
2'-Hydroxy-5'-methylacetophenone 98%
2'-Hydroxy-5'-methylacetophenone
Purity: 98%
CAS No.: 1450-72-2
Molecular Formula: C9H10O2
Formula Weight: 150.18
Flash Point:
Formula:
C₉H₁₀O₂ Boiling Pt: 112…114 °C/12 mm Melting Pt: 45…48 °C Density: 1,079 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002380 CAS Number: 1450-72-2 EINECS: 215-915-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...