2-Bromo-m-xylene 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13130-14
A13130-22
A13130-36
A13130-09
A13130-30
CAAAAA13130-14EA
145.99
CAD
CAAAAA13130-14
CAAAAA13130-22
CAAAAA13130-36
CAAAA13130-09
CAAAA13130-30
2-Bromo-m-xylene 98+%
2-Bromo-m-xylene
Purity: 98+%
CAS No.: 576-22-7
Molecular Formula: C8H9Br
Formula Weight: 185.07
Flash Point: 73°(163°F)
Refractive Index: 1.5550
Melting Point: -10°
Boiling Point: 205-206°
Beilstein Reference: 1929780
MDL No.: MFCD00000075
Formula:
C₈H₉Br Boiling Pt: 205…206 °C Melting Pt: –10 °C Density: 1.384 Flash Pt: 73 °C(163 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000075 CAS Number: 576-22-7 EINECS: 209-397-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...