α-Bromo-2-nitrotoluene 98+%

Supplier: Thermo Scientific Chemicals
Danger

A13127-06 A13127-14 A13127-22
CAAAA13127-06EA 89.27 CAD
CAAAA13127-06 CAAAA13127-14 CAAAA13127-22
α-Bromo-2-nitrotoluene 98+%
α-Bromo-2-nitrotoluene

Purity: 98+% CAS No.: 3958-60-9 Molecular Formula: C7H6BrNO2 Formula Weight: 216.04 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 45-48° Beilstein Reference: 638991 UN No.: 3261 MDL No.: MFCD00007184

Formula: C₇H₆BrNO₂
Melting Pt: 45…48 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00007184
CAS Number: 3958-60-9
EINECS: 223-558-9
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR