5-Chloroisatin 98%

Supplier: Thermo Scientific Chemicals

A13125-06 A13125-14 A13125-22
CAAAA13125-06EA 40.92 CAD
CAAAA13125-06 CAAAA13125-14 CAAAA13125-22
5-Chloroisatin 98%
5-Chloroisatin

Purity: 98% CAS No.: 17630-76-1 Molecular Formula: C8H4ClNO2 Formula Weight: 181.58 Melting Point: ca 247° dec. Beilstein Reference: 383759

Formula: C₈H₄ClNO₂
Boiling Pt: 361 °C (1013 hPa)
Melting Pt: ∼ 247 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00014567
CAS Number: 17630-76-1
EINECS: 241-614-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR