1-Chloro-3-fluorobenzene 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13123-36
A13123-14
A13123-22
CAAAAA13123-36EA
878.48
CAD
CAAAAA13123-36
CAAAA13123-14
CAAAA13123-22
1-Chloro-3-fluorobenzene 99%
1-Chloro-3-fluorobenzene
Purity: 99%
CAS No.: 625-98-9
Molecular Formula: C6H4ClF
Formula Weight: 130.55
Flash Point: 20°(68°F)
Refractive Index: 1.4940
Boiling Point: 126-128°
Beilstein Reference: 2039303
UN No.: 1993
MDL No.: MFCD00000569
Formula:
C₆H₄ClF Boiling Pt: 126…128 °C Density: 1.224 Flash Pt: 20 °C(68 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000569 CAS Number: 625-98-9 EINECS: 210-919-0 UN: 1993 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...