1H-1,2,4-Triazole-3-thiol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13120-06
A13120-14
A13120-22
CAAAA13120-06EA
67.62
CAD
CAAAA13120-06
CAAAA13120-14
CAAAA13120-22
1H-1,2,4-Triazole-3-thiol 98%
1H-1,2,4-Triazole-3-thiol
Purity: 98%
CAS No.: 3179-31-5
Molecular Formula: C2H3N3S
Formula Weight: 101.13
Melting Point: 217-224°
Beilstein Reference: 107731
Formula:
C₂H₃N₃S Melting Pt: 217…224 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005229 CAS Number: 3179-31-5 EINECS: 221-656-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...