3-Methylquinoline 98+%

Supplier: Thermo Scientific Chemicals
Warning

A13109-06 A13109-14
CAAAAA13109-22EA 3239.36 CAD
CAAAAA13109-22 CAAAA13109-06 CAAAA13109-14
3-Methylquinoline 98+%
3-Methylquinoline

Purity: 98+% CAS No.: 612-58-8 Molecular Formula: C10H9N Formula Weight: 143.19 Flash Point: 110°(230°F) Refractive Index: 1.6150 Melting Point: 15° Boiling Point: 252-253° Beilstein Reference: 110325 MDL No.: MFCD00014661

Formula: C₁₀H₉N
MW: 143.19 g/mol
Boiling Pt: 252…253 °C
Melting Pt: 15 °C
Density: 1.069
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014661
CAS Number: 612-58-8
EINECS: 210-315-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR