Ethyl pivalate 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trimethylacetic acid ethyl ester, Ethyltrimethylacetate, Pivalic acid ethyl ester, 2,2-Dimethylpropionic acid ethyl ester, Ethyl trimethylacetate

A13100-14 A13100-22
CAAAA13100-14EA 49.45 CAD
CAAAA13100-14 CAAAA13100-22 CAAAA13100-36
Ethyl pivalate 99%
Ethyl pivalate

Purity: 99% CAS No.: 3938-95-2 Molecular Formula: (CH3)3CCO2CH2CH3 Formula Weight: 130.19 Flash Point: 16°(60°F) Refractive Index: 1.3930 Odor: Stench Melting Point: -90° Boiling Point: 116-118° Beilstein Reference: 1747617 Merck Reference: 14,7511 UN No.: 3272 MDL No.: MFCD00009129

Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 118
Melting Pt: -90
Density: 0.891
Flash Pt: 16 °C (60 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009129
CAS Number: 3938-95-2
EINECS: 223-520-1
UN: 3272
ADR: 3 III
Merck Index: 14,7511

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR