2-Mercapto-1-methylimidazole 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13094-14
A13094-22
CAAAA13094-14EA
87.85
CAD
CAAAA13094-14
CAAAA13094-22
2-Mercapto-1-methylimidazole 98%
2-Mercapto-1-methylimidazole
Possesses antioxidant properties
Purity: 98%
CAS No.: 60-56-0
Molecular Formula: C4H6N2S
Formula Weight: 114.17
Odor: Stench
Melting Point: 144-147°
Boiling Point: 280° dec.
Solubility: Freely soluble in water. Soluble in alcohol, chloroform. Sparingly soluble in ether, benzene.
Beilstein Reference: 108646
Merck Reference: 14,5971
MDL No.: MFCD00179321
Formula:
C₄H₆N₂S Boiling Pt: 280 °C (dec.) Melting Pt: 144…147 °C Density: 1.36 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00179321 CAS Number: 60-56-0 EINECS: 200-482-4 UN: 3335 ADR: 9, Merck Index: 14,5971 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...