1-Pentanol 98+%

Supplier: Thermo Scientific Chemicals
Warning

A13093-0D A13093-0F A13093-AP
CAAAA13093-0DEA 497.75 CAD
CAAAA13093-0D CAAAA13093-0F CAAAA13093-AP
1-Pentanol 98+%
1-Pentanol

Purity: 98+% CAS No.: 71-41-0 Molecular Formula: C5H12O Formula Weight: 88.15 Flash Point: 49°(120°F) Refractive Index: 1.4103 Odor: Mild Melting Point: -79° Boiling Point: 136-138° Beilstein Reference: 1730975 Merck Reference: 14,7118 UN No.: 1105 MDL No.: MFCD00002977

Formula: H₃C(CH₂)₃CH₂OH
Boiling Pt: 136…138 °C
Melting Pt: –79 °C
Density: 0.812
Flash Pt: 49 °C (120 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002977
CAS Number: 71-41-0
EINECS: 200-752-1
UN: 1105
ADR: 3,III
Merck Index: 14,7118
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92dac1192eb8e068fc1804fdcf488129a2385918&vid=d4c7f2c5672a97c6dfaddc50807d768068096252&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR