4-Bromo-2-fluorobenzonitrile 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Brom-4-cyan-3-fluorbenzene

A13086-06 A13086-14
CAAAAA13086-06EA 56.88 CAD
CAAAAA13086-06 CAAAA13086-03 CAAAA13086-14
4-Bromo-2-fluorobenzonitrile 99%
4-Bromo-2-fluorobenzonitrile

Purity: 99% CAS No.: 105942-08-3 Molecular Formula: C7H3BrFN Formula Weight: 200.01 Melting Point: 69-72° Boiling Point: 238-239° UN No.: 3439 MDL No.: MFCD00143264

Formula: C₇H₃BrFN
MW: 200.01 g/mol
Boiling Pt: 238…239 °C
Melting Pt: 69…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00143264
CAS Number: 105942-08-3
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR