2-(2-Chloroethoxy)ethanol 99%

Supplier: Thermo Scientific Chemicals
Warning

A13074-0B A13074-18 A13074-30
CAAAA13074-0BEA 843.72 CAD
CAAAA13074-0B CAAAA13074-18 CAAAA13074-30
2-(2-Chloroethoxy)ethanol 99%
2-(2-Chloroethoxy)ethanol

Purity: 99% CAS No.: 628-89-7 Molecular Formula: C4H9ClO2 Formula Weight: 124.57 Flash Point: 90°(194°F) Refractive Index: 1.4535 Boiling Point: 185° Beilstein Reference: 506015 MDL No.: MFCD00002870

Formula: C₄H₉ClO₂
MW: 124.57 g/mol
Boiling Pt: 185 °C
Density: 1.180
Flash Pt: 90 °C (194 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002870
CAS Number: 628-89-7
EINECS: 211-059-9
UN: 3334
ADR: 9,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR