1,2,4-Trihydroxybenzene 97%

Supplier: Thermo Scientific Chemicals
Warning

A13069-03 A13069-06 A13069-14
CAAAAA13069-03EA 69.84 CAD
CAAAAA13069-03 CAAAA13069-06 CAAAA13069-14
1,2,4-Trihydroxybenzene 97%
1,2,4-Trihydroxybenzene

Purity: 97% CAS No.: 533-73-3 Molecular Formula: C6H6O3 Formula Weight: 126.11 Melting Point: ca 140° subl. Beilstein Reference: 2042863 Merck Reference: 14,1073 MDL No.: MFCD00002198

Formula: C₆H₆O₃
Boiling Pt: 362 °C (1013 hPa)
Melting Pt: ∼ 140 °C (subl.)
Density: 1.45 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002198
CAS Number: 533-73-3
EINECS: 208-575-1
Merck Index: 14,1073

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR