(4-Chlorophenoxy)acetic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13065-0E
A13065-36
CAAAA13065-0EEA
511.92
CAD
CAAAA13065-0E
CAAAA13065-36
(4-Chlorophenoxy)acetic acid 98+%
(4-Chlorophenoxy)acetic acid
Purity: 98+%
CAS No.: 122-88-3
Molecular Formula: C8H7ClO3
Formula Weight: 186.59
Melting Point: 156-161°
Beilstein Reference: 1211804
Formula:
(C₆H₄Cl)OCH₂COOH Boiling Pt: 308 °C (1013 hPa) Melting Pt: 156…161 °C Density: 1.53 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004305 CAS Number: 122-88-3 EINECS: 204-581-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...