tert-Butylchlorodimethylsilane 97%

Supplier: Thermo Scientific Chemicals
Danger

A13064-06 A13064-14 A13064-22
CAAAA13064-06EA 51.35 CAD
CAAAA13064-06 CAAAA13064-14 CAAAA13064-22
tert-Butylchlorodimethylsilane 97%
tert-Butylchlorodimethylsilane

Purity: 97% CAS No.: 18162-48-6 Molecular Formula: C6H15ClSi Formula Weight: 150.73 Flash Point: 22°(72°F) Odor: Pungent Boiling Point: 124-126° Beilstein Reference: 505999 UN No.: 2925 MDL No.: MFCD00000501

Formula: C₆H₁₅ClSi
Boiling Pt: 124…126 °C
Melting Pt: 86…89 °C
Density: 0.81
Flash Pt: 22 °C (72 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000501
CAS Number: 18162-48-6
EINECS: 242-042-4
UN: 2921
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR