2-Acetylthiophene 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13062-22
A13062-36
CAAAA13062-22EA
85.64
CAD
CAAAA13062-22
CAAAA13062-36
2-Acetylthiophene 99%
2-Acetylthiophene
Purity: 99%
CAS No.: 88-15-3
Molecular Formula: C6H6OS
Formula Weight: 126.18
Flash Point: 91°(195°F)
Refractive Index: 1.5660
Odor: Slightly sweetish
Melting Point: 10-12°
Boiling Point: 213-214°
Beilstein Reference: 107910
UN No.: 2810
MDL No.: MFCD00005442
Formula:
C₆H₆OS Boiling Pt: 213…214 °C Melting Pt: 10…12 °C Density: 1.168 Flash Pt: 91 °C (195 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005442 CAS Number: 88-15-3 EINECS: 201-804-6 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...