2-Iodoaniline 98+%

Supplier: Thermo Scientific Chemicals
Warning

A13059-06 A13059-14 A13059-22 A13059-18
CAAAAA13059-06EA 69.99 CAD
CAAAAA13059-06 CAAAAA13059-14 CAAAAA13059-22 CAAAA13059-18
2-Iodoaniline 98+%
2-Iodoaniline

Purity: 98+% CAS No.: 615-43-0 Molecular Formula: C6H6IN Formula Weight: 219.03 Flash Point: 110°(230°F) Melting Point: 53-57° Beilstein Reference: 2204899 MDL No.: MFCD00007680

Formula: C₆H₆IN
MW: 219.02 g/mol
Boiling Pt: 263 °C (1013 hPa)
Melting Pt: 53…57 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007680
CAS Number: 615-43-0
EINECS: 210-426-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR