Ethyl benzoate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13057-0E
A13057-22
A13057-36
CAAAA13057-0EEA
202.24
CAD
CAAAA13057-0E
CAAAA13057-22
CAAAA13057-36
Ethyl benzoate 99%
Ethyl benzoate
Purity: 99%
CAS No.: 93-89-0
Molecular Formula: C9H10O2
Formula Weight: 150.18
Flash Point: 84°(183°F)
Refractive Index: 1.5050
Melting Point: -34°
Boiling Point: 212-214°
Beilstein Reference: 1908172
Merck Reference: 14,3766
MDL No.: MFCD00009109
Formula:
C₉H₁₀O₂ Boiling Pt: 212…214 °C Melting Pt: –34 °C Density: 1.049 Flash Pt: 84 °C(183 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009109 CAS Number: 93-89-0 EINECS: 202-284-3 Merck Index: 14,3766 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...