Ethyl-3-phenylpropionate 98+%
Supplier: Thermo Scientific Chemicals
A13055-0B
A13055-18
A13055-30
CAAAAA13055-0BEA
1188.16
CAD
CAAAAA13055-0B
CAAAA13055-18
CAAAA13055-30
Ethyl-3-phenylpropionate 98+%
Ethyl 3-phenylpropionate
Purity: 98+%
CAS No.: 2021-28-5
Molecular Formula: C6H5CH2CH2CO2CH2CH3
Formula Weight: 178.23
Flash Point: 107°(224°F)
Refractive Index: 1.4940
Boiling Point: 118°/3mm
Beilstein Reference: 1910432
MDL No.: MFCD00009206
Formula:
C₁₁H₁₄O₂ Boiling Pt: 118 °C/3 mm Density: 1.016 Flash Pt: 107 °C(224 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009206 CAS Number: 2021-28-5 EINECS: 217-966-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...