2-Aminopyrazine 99%

Supplier: Thermo Scientific Chemicals
Warning

A13052-06 A13052-14 A13052-18
CAAAAA13052-06EA 85.95 CAD
CAAAAA13052-06 CAAAAA13052-14 CAAAA13052-18
2-Aminopyrazine 99%
2-Aminopyrazine

Purity: 99% CAS No.: 5049-61-6 Molecular Formula: C4H5N3 Formula Weight: 95.11 Melting Point: 118-121° Beilstein Reference: 107025

Formula: C₄H₅N₃
Boiling Pt: 203 °C (1013 hPa)
Melting Pt: 118…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00006137
CAS Number: 5049-61-6
EINECS: 225-748-7
UN: 1325
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR